MilliporeSigma
  • Prostaglandin E2 Promotes Colorectal Cancer Stem Cell Expansion and Metastasis in Mice.
Prostaglandin E2 Promotes Colorectal Cancer Stem Cell Expansion and Metastasis in Mice.
Gastroenterology (2015-08-12)
Dingzhi Wang, Lingchen Fu, Haiyan Sun, Lixia Guo, Raymond N DuBois
ABSTRACT
Inflammation may contribute to the formation, maintenance, and expansion of cancer stem cells (CSCs), which have the capacity for self-renewal, differentiation, and resistance to cytotoxic agents. We investigated the effects of the inflammatory mediator prostaglandin E2 (PGE2) on colorectal CSC development and metastasis in mice and the correlation between levels of PGE2 and CSC markers in human colorectal cancer (CRC) specimens. Colorectal carcinoma specimens and matched normal tissues were collected from patients at the Mayo Clinic (Scottsdale, AZ) and analyzed by mass spectrometry and quantitative polymerase chain reaction. Human primary CRC cells and mouse tumor cells were isolated using microbeads or flow cytometry and analyzed for sphere-formation and by flow cytometry assays. LS-174T cells were sorted by flow cytometry (for CD133(+)CD44(+) and CD133(-)CD44(-) cells) and also used in these assays. NOD-scidIL-2Rγ(-/-) (NSG) mice were given cecal or subcutaneous injections of LS-174T or human primary CRC cells. Apc(Min/+) mice and NSG mice with orthotopic cecal tumors were given vehicle (controls), PGE2, celecoxib, and/or Ono-AE3-208. PGE2 downstream signaling pathways were knocked down with small hairpin RNAs, expressed from lentiviral vectors in LS-174T cells, or blocked with inhibitors in human primary CRC cells. Levels of PGE2 correlated with colonic CSC markers (CD133, CD44, LRG5, and SOX2 messenger RNAs) in human colorectal carcinoma samples. Administration of PGE2 to Apc(Min/+) mice increased tumor stem cells and tumor burden, compared with controls. NSG mice given PGE2 had increased numbers of cecal CSCs and liver metastases compared with controls after intracecal injection of LS-174T or human primary CRC cells. Alternatively, celecoxib, an inhibitor of prostaglandin-endoperoxide synthase 2, reduced polyp numbers in Apc(Min/+) mice, liver metastasis in NSG mice with orthotopic tumors, and numbers of CSCs in Apc(Min/+) and NSG mice. Inhibitors or knockdown of PGE2 receptor 4 (EP4), phosphoinositide 3-kinase (PI3K) p85α, extracellular signal-regulated kinase 1 (ERK1), or nuclear factor (NF)-κB reduced PGE2-induced sphere formation and expansion of LS-174T and/or human primary CRC cells. Knockdown of ERK1 or PI3K p85α also attenuated PGE2-induced activation of NF-κB in LS-174T cells. An EP4 antagonist reduced the ability of PGE2 to induce CSC expansion in orthotopic tumors and to accelerate the formation of liver metastases. Knockdown experiments showed that NF-κB was required for PGE2 induction of CSCs and metastasis in mice. PGE2 induces CSC expansion by activating NF-κB, via EP4-PI3K and EP4-mitogen-activated protein kinase signaling, and promotes the formation of liver metastases in mice. The PGE2 signaling pathway therefore might be targeted therapeutically to slow CSC expansion and colorectal cancer progression.
Sigma-Aldrich
MISSION® esiRNA, targeting human PTGER4
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ptger4
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
PD 98,059, solid
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)