MilliporeSigma
  • Effects of Plant-Derived Extracts, Other Antimicrobials, and Their Combinations against Escherichia coli O157:H7 in Beef Systems.
Effects of Plant-Derived Extracts, Other Antimicrobials, and Their Combinations against Escherichia coli O157:H7 in Beef Systems.
Journal of food protection (2015-06-04)
Kyung Yuk Ko, Ifigenia Geornaras, Hyun-Dong Paik, Kee-Tae Kim, John N Sofos
ABSTRACT
The antimicrobial effects of thyme oil (TO), grapefruit seed extract (GSE), and basil essential oil, alone or in combination with cetylpyridinium chloride (CPC), sodium diacetate, or lactic acid, were evaluated against Escherichia coli O157:H7 in a moisture-enhanced beef model system. The model system was composed of a nonsterile beef homogenate to which NaCl (0.5%) and sodium tripolyphosphate (0.25%) were added, together with the tested antimicrobial ingredients. Beef homogenate treatments were inoculated (ca. 3 log CFU/ml) with rifampin-resistant E. coli O157:H7 (eight-strain mixture) and incubated at 15 °C (48 h). The most effective individual treatments were TO (0.25 or 0.5%) and GSE (0.5 or 1.0%), which immediately reduced (P < 0.05) pathogen levels by ≥ 3.4 log CFU/ml. Additionally, CPC (0.04%) reduced initial E. coli O157:H7 counts by 2.7 log CFU/ml. Most combinations of the tested plant-derived extracts with CPC (0.02 or 0.04%) and sodium diacetate (0.25%) had an additive effect with respect to antibacterial activity. In a second study, antimicrobial interventions were evaluated for their efficacy in reducing surface contamination of E. coli O157:H7 on beef cuts and to determine the effect of these surface treatments on subsequent internalization of the pathogen during blade tenderization. Beef cuts (10 by 8 by 3.5 cm) were inoculated (ca. 4 log CFU/g) on one side with the rifampin-resistant E. coli O157:H7 strain mixture and were then spray treated (20 lb/in(2), 10 s) with water, GSE (5 and 10%), lactic acid (5%), or CPC (5%). Untreated (control) and spray-treated surfaces were then subjected to double-pass blade tenderization. Surface contamination (4.4 log CFU/g) of E. coli O157:H7 was reduced (P < 0.05) to 3.4 (5% CPC) to 4.1 (water or 5% GSE) log CFU/g following spray treatment. The highest and lowest transfer rates of pathogen cells from the surface to deeper tissues of blade-tenderized sections were obtained in the untreated control and CPC-treated samples, respectively.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Cetylpyridinium chloride, meets USP testing specifications
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, for molecular biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, for molecular biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis