MilliporeSigma
  • Roundabout4 suppresses glioma-induced endothelial cell proliferation, migration and tube formation in vitro by inhibiting VEGR2-mediated PI3K/AKT and FAK signaling pathways.
Roundabout4 suppresses glioma-induced endothelial cell proliferation, migration and tube formation in vitro by inhibiting VEGR2-mediated PI3K/AKT and FAK signaling pathways.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-04-04)
Heng Cai, Yixue Xue, Zhen Li, Yi Hu, Zhenhua Wang, Wenjing Liu, Zhiqing Li, Yunhui Liu
ABSTRACT
Endothelial cell (EC) proliferation, migration, and tube formation are the critical steps for tumor angiogenesis, which is involved in the formation of new tumor blood vessels. Roundabout4 (Robo4), a new member of Robo proteins family, is specifically expressed in endothelial cells. This study aimed to investigate the effects of Robo4 on glioma-induced endothelial cell proliferation, migration and tube formation in vitro. We found that Robo4 was endogenously expressed in Human Brain Microvascular Endothelial Cells (HBMECs), while Robo4 was significantly down-regulated in endothelial cells cultured in glioma conditioned medium. Robo4 over-expression remarkably suppressed glioma-induced endothelial cell proliferation, migration and tube formation in vitro. In addition, Robo4 influenced the glioma-induced angiogenesis via binding to its ligand Slit2. Further studies demonstrated that the knockdown of Robo4 up-regulated the phosphorylation of VEGFR2, PI3K, AKT and FAK in EC cultured in glioma conditioned medium. VEGFR2 inhibitor SU-1498, AKT inhibitor LY294002 and FAK inhibitor 14 (FAK inhibitor) blocked the Robo4 knockdown-mediated alteration in glioma angiogenesis in vitro. Our results proved that Robo4 suppressed glioma-induced endothelial cell proliferation, migration and tube formation in vitro by inhibiting VEGR2-mediated activation of PI3K/AKT and FAK signaling pathways.
Sigma-Aldrich
MISSION® esiRNA, targeting human ROBO4
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)