MilliporeSigma
  • Epigenetic modification and preliminary investigation of the mechanism of the immune evasion of HL-60 cells.
Epigenetic modification and preliminary investigation of the mechanism of the immune evasion of HL-60 cells.
Molecular medicine reports (2015-03-31)
Jin Hong Liu, Yong Mei Bian, Yi Xie, Dao Pei Lu
ABSTRACT
The aim of the present study was to explore the effect of epigenetic modification of class II transactivator (CIITA) methylation on histocompatibility complex (MHC) class II expression and the immune evasion of leukemia HL-60 cells. HL-60 cells were treated with various concentrations of 5-aza-2'deoxycytidine (5-Aza-CdR) and 0.5 µmol/l suberoylanilide hydroxamic acid (SAHA) for 24 h and then stimulated by interferon γ (IFN-γ) for 48 h. The mRNA levels of MHC class I, II and co-stimulatory molecules were quantified by reverse transcription polymerase chain reaction (RT-PCR). The levels of CIITA protein were determined by western blot analysis, and the CpG island methylation ratios in the CIITA promoter IV (CIITApIV) were analyzed by bisulfite-sequencing PCR (BSP). MHC I as well as the co-stimulatory molecules CD40 and CD80 were significantly increased following treatment with 5-Aza-CdR + SAHA + IFN-γ (epigenetic groups) compared with those in the control group and IFN-γ group (P<0.05). The expression of MHC class II and CIITA was restored and increased in an 5-Aza-CdR concentration-dependent manner in the three epigenetic groups. The results of the BSP assay showed that the methylation rate of CIITApIV CpG sites decreased with the treatment of epigenetic modification and negatively correlated to the 5-Aza-CdR concentration. This demonstrated that the negative expression of CIITA protein was the key reason for the loss of MHC II expression in HL-60 cells. The results of the present study may help to illustrate the mechanism of immune evasion in HL-60 cells.
Sigma-Aldrich
Ethidium bromide solution, for fluorescence, ~1% in H2O
Supelco
Pyridaben, PESTANAL®, analytical standard
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 500 μg/mL in H2O
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Ethidium bromide solution, BioReagent, for molecular biology, 10 mg/mL in H2O
Sigma-Aldrich
Laemmli Lysis-buffer, non smelling
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Ethidium bromide, BioReagent, for molecular biology, powder
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ethidium bromide, ~95% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
SAHA, ≥98% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, Vetec, reagent grade, 99%