MilliporeSigma
  • Nickel-catalysed cyclopropanation of electron-deficient alkenes with diiodomethane and diethylzinc.
Nickel-catalysed cyclopropanation of electron-deficient alkenes with diiodomethane and diethylzinc.
Chemical Communications (Cambridge, England) (2016)
Xu J, et al.