MilliporeSigma
  • Whole-mount three-dimensional imaging of internally localized immunostained cells within mouse embryos.
Whole-mount three-dimensional imaging of internally localized immunostained cells within mouse embryos.
Nature protocols (2012-02-11)
Tomomasa Yokomizo, Tomoko Yamada-Inagawa, Amanda D Yzaguirre, Michael J Chen, Nancy A Speck, Elaine Dzierzak
ABSTRACT
We describe a three-dimensional (3D) confocal imaging technique to characterize and enumerate rare, newly emerging hematopoietic cells located within the vasculature of whole-mount preparations of mouse embryos. However, the methodology is broadly applicable for examining the development and 3D architecture of other tissues. Previously, direct whole-mount imaging has been limited to external tissue layers owing to poor laser penetration of dense, opaque tissue. Our whole-embryo imaging method enables detailed quantitative and qualitative analysis of cells within the dorsal aorta of embryonic day (E) 10.5-11.5 embryos after the removal of only the head and body walls. In this protocol we describe the whole-mount fixation and multimarker staining procedure, the tissue transparency treatment, microscopy and the analysis of resulting images. A typical two-color staining experiment can be performed and analyzed in ∼6 d.
Sigma-Aldrich
Benzyl benzoate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Benzyl alcohol, ACS reagent, ≥99.0%
Sigma-Aldrich
Anti-phospho-Histone H3 (Ser10) Antibody, clone 3H10, clone 3H10, Upstate®, from mouse