MilliporeSigma
  • B7-H3 silencing inhibits tumor progression of mantle cell lymphoma and enhances chemosensitivity.
B7-H3 silencing inhibits tumor progression of mantle cell lymphoma and enhances chemosensitivity.
International journal of oncology (2015-04-16)
Wei Zhang, Yanfang Wang, Jing Wang, Fei Dong, Mingxia Zhu, Wenli Wan, Haishen Li, Feifei Wu, Xinxing Yan, Xiaoyan Ke
ABSTRACT
B7-H3 (CD276), known as a member of B7 immunoregulatory family, is a type I transmembrane glycoprotein aberrantly expressed in numerous types of cancer and associated with poor prognosis. However, the role of B7-H3 in oncogenesis and chemosensitivity of mantle cell lymphoma (MCL) remains unknown. We determined the effects of downregulating B7-H3 expression on tumor progression and the sensitivity of chemotherapeutic drug in mantle cell lymphoma. B7-H3 knockdown was performed using lentivirus transduction in the Maver and Z138 mantle cell lymphoma cell lines, respectively. The effects of B7-H3 on cell proliferation, cycle, migration and invasion were investigated by CCK-8 assay, methyl cellulose colony forming assay, PI staining, and Transwell assays in vitro. By establishing Maver and Z138 xenograft models, the effects of B7-H3 on tumorigenicity were observed, and Ki-67 and PCNA was detected by immunohistochemistry. The downregulation of B7-H3 significantly decreased tumor proliferation in MCL in vitro and in vivo. In the B7-H3 knockdown groups of Maver and Z138 xenograft models, the mean inhibition rate of tumor growth was 59.1 and 65.0% (p=0.010 and 0.003), and the expression of both Ki-67 and PCNA were significantly lower, respectively. After B7-H3 silencing, the cell cycles of Maver and Z138 were both arrested at G0/G1 phase, and the cell migration rates and invasion capacity were decreased as well. Moreover, the impacts of B7-H3 RNAi on the antitumor effect of chemotherapy drugs were determined with CCK-8 and Annexin V-FITC/PI assays in vitro and with xenograft models in vivo. The silencing of B7-H3 increased the sensitivity of Maver and Z138 cells to rituximab and bendamustine and enhanced the drug-induced apoptosis, respectively. Our study demonstrates for the first time that B7-H3 promotes mantle cell lymphoma progression and B7-H3 knockdown significantly enhances the chemosensitivity. This may provide a new therapeutic approach to mantle cell lymphoma.
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Anti-CD276 antibody produced in rabbit, Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
SAFC
L-Glutamine
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture