MilliporeSigma
  • 2,2,6,6-Tetramethylpiperidine-catalyzed, ortho-selective chlorination of phenols by sulfuryl chloride.
2,2,6,6-Tetramethylpiperidine-catalyzed, ortho-selective chlorination of phenols by sulfuryl chloride.
The Journal of organic chemistry (2013-12-18)
Noam I Saper, Barry B Snider
ABSTRACT
2,2,6,6-Tetramethylpiperidine (TMP)-catalyzed (1-10%) chlorinations of phenols by SO2Cl2 in aromatic solvents are more ortho selective than with primary and less hindered secondary amine catalysts. Ortho-selective chlorination is successful even with electron deficient phenols such as 2-hydroxybenzaldehyde and 2'-hydroxyacetophenone. Notably, ortho selectivity increases with the reaction temperature. On the other hand, tetraalkylammonium chloride-catalyzed chlorinations are moderately para selective.
Sigma-Aldrich
Sulfuryl chloride, 97%