MilliporeSigma
  • Quantitative microscopy of functional HIV post-entry complexes reveals association of replication with the viral capsid.
Quantitative microscopy of functional HIV post-entry complexes reveals association of replication with the viral capsid.
eLife (2014-12-18)
Ke Peng, Walter Muranyi, Bärbel Glass, Vibor Laketa, Stephen R Yant, Luong Tsai, Tomas Cihlar, Barbara Müller, Hans-Georg Kräusslich
ABSTRACT
The steps from HIV-1 cytoplasmic entry until integration of the reverse transcribed genome are currently enigmatic. They occur in ill-defined reverse-transcription- and pre-integration-complexes (RTC, PIC) with various host and viral proteins implicated. In this study, we report quantitative detection of functional RTC/PIC by labeling nascent DNA combined with detection of viral integrase. We show that the viral CA (capsid) protein remains associated with cytoplasmic RTC/PIC but is lost on nuclear PIC in a HeLa-derived cell line. In contrast, nuclear PIC were almost always CA-positive in primary human macrophages, indicating nuclear import of capsids or capsid-like structures. We further show that the CA-targeted inhibitor PF74 exhibits a bimodal mechanism, blocking RTC/PIC association with the host factor CPSF6 and nuclear entry at low, and abrogating reverse transcription at high concentrations. The newly developed system is ideally suited for studying retroviral post-entry events and the roles of host factors including DNA sensors and signaling molecules.
Sigma-Aldrich
Anti-CPSF6 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution