MilliporeSigma
  • Loss of ARID1A/BAF250a expression in ovarian endometriosis and clear cell carcinoma.
Loss of ARID1A/BAF250a expression in ovarian endometriosis and clear cell carcinoma.
International journal of clinical and experimental pathology (2012-09-15)
Wenbin Xiao, Amad Awadallah, Wei Xin
ABSTRACT
Ovarian endometriosis has been associated with increased risk for ovarian clear cell carcinoma (CCC). Atypical endometriosis shares common molecular alterations with CCC and therefore, has been proposed as a precursor lesion of CCC, although it is unclear if benign endometriosis is pre-neoplastic. In this study, we examined some molecular alterations in ovarian benign endometriosis, atypical endometriosis, and CCC in comparison to papillary serous carcinoma (PSC). These included BAF250a (encoded by ARID1A), a recently identified major tumor suppressor in ovarian CCC, as well as hepatocyte nuclear factor (HNF)-1b, estrogen receptor (ER), progesterone receptor (PR), and P53. We confirmed that CCC but not PSC had loss of BAF250a expression, HNF-1b up-regulation, loss of ER expression and P53 expression. We further showed that both atypical endometriosis and adjacent CCC had loss of BAF250a expression (38.5% vs. 57.7%), HNF-1b up-regulation (53.8% vs. 92.3%), and loss of ER (84.6% vs. 92.3%) and PR (76.9% vs. 84.6%) expression. Importantly, about 20% of benign ovarian endometriosis had loss of BAF250a expression, 33% with HNF-1b up-regulation, 23% loss of ER expression and 50% loss of PR expression, respectively. The concurrent rate of loss of BAF250a expression, HNF-1b up-regulation, and loss of ER expression was not observed in any benign endometriosis, and was increased to 23.1% in atypical endometriosis, and was further increased to 42.3% in CCC. Therefore, the molecular alterations accumulate in a stepwise manner along the transformation process from benign endometriosis through atypical endometriosis to CCC. These data suggest that a portion of benign ovarian endometriosis has already undergone genetic alterations that lead to aberrant protein expression, possibly conferring a higher risk for malignant transformation.
Sigma-Aldrich
Anti-ARID1A antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution