MilliporeSigma
  • Specificity of T lymphocyte cytotoxicity to autologous hepatocytes in chronic hepatitis B virus infection: evidence that T cells are directed against HBV core antigen expressed on hepatocytes.
Specificity of T lymphocyte cytotoxicity to autologous hepatocytes in chronic hepatitis B virus infection: evidence that T cells are directed against HBV core antigen expressed on hepatocytes.
Journal of immunology (Baltimore, Md. : 1950) (1982-12-01)
M Mondelli, G M Vergani, A Alberti, D Vergani, B Portmann, A L Eddleston, R Williams
ABSTRACT
Peripheral blood T lymphocytes from 21 patients with chronic HBV infection were incubated with autologous hepatocytes in a microcytotoxicity assay. Cytotoxicity was significantly increased in 13 cases, and in 12 of these the cytotoxic effect of the T lymphocytes was inhibited by preincubating the liver cells with IgG containing antibodies to the hepatitis B core antigen (HBcAg). Normal human IgG and IgG containing antibodies to the hepatitis B surface antigen (HBsAG) were without effect. Control experiments using autologous fibroblasts as target cells showed low levels of T cell cytotoxicity and no blocking effect of anti-core antibody. All patients in whom it was possible to demonstrate HBcAg in liver tissue had significantly increased T cell cytotoxicity to autologous hepatocytes. These studies suggest that T cell cytotoxicity in patients with chronic HBV infection is directed against determinants resembling the hepatitis B core antigen on the plasma membrane of hepatocytes.
Sigma-Aldrich
HBcAg core human, ~1 mg/mL, ≥95% (SDS-PAGE), recombinant, expressed in E. coli