MilliporeSigma
  • Functional role of the extended loop 2 in the myosin 9b head for binding F-actin.
Functional role of the extended loop 2 in the myosin 9b head for binding F-actin.
The Journal of biological chemistry (2008-12-09)
Sandra Struchholz, Kerstin Elfrink, Uwe Pieper, Georg Kalhammer, Ulrike Honnert, Anika Grützner, Wolfgang A Linke, Wanqin Liao, Martin Bähler
ABSTRACT
The mammalian class IX myosin Myo9b can move considerable distances along actin filaments before it dissociates. This is remarkable, because it is single headed and because the rate-limiting step in its ATPase cycle is ATP hydrolysis. Thus, it spends most of its cycling time in the ATP-bound state that has a weak affinity for F-actin in other myosins. It has been speculated that the very extended loop 2 in the Myo9b head domain comprises an additional actin-binding site that prevents it from dissociation in the weak binding states. Here we show that two regions in the loop 2 determine the F-actin concentrations needed to maximally activate the steady-state ATPase activity. Together these two regions regulate the amount capable of binding F-actin and the affinity of the nucleotide-free state. The isolated loop 2 behaved like an entropic spring and bound stoichiometrically and with high affinity to F-actin. Subfragment 1 from skeletal muscle myosin II bound to F-actin simultaneously with the isolated loop 2 of Myo9b and could not displace it. Furthermore, the present results imply also a regulatory role for the tail region. Taken together, the results demonstrate that the extended loop 2 in Myo9b binds F-actin and influences the binding of the conventional stereo-specific actin-binding site.