MilliporeSigma
  • NKX3.1 is expressed in ER-positive and AR-positive primary breast carcinomas.
NKX3.1 is expressed in ER-positive and AR-positive primary breast carcinomas.
Journal of clinical pathology (2014-07-06)
Rebecca J Asch-Kendrick, Mark A Samols, Mohammed T Lilo, Andrea P Subhawong, Rajni Sharma, Peter B Illei, Pedram Argani, Ashley Cimino-Mathews
ABSTRACT
NKX3.1 is an androgen-regulated tumour suppressor gene that is downregulated in prostate carcinoma. Immunohistochemistry for NKX3.1 is primarily specific for prostatic-derived tumours and tissue but is reported in a small number of breast carcinomas. NKX3.1 is also shown to inhibit estrogen receptor (ER) signalling in breast carcinoma models. Here, we investigate labelling of NKX3.1 in invasive ductal (IDC) and lobular (ILC) carcinomas of the breast with full characterisation of ER, progesterone receptor (PR), androgen receptor (AR) and Her2 status. Tissue microarrays of 86 primary IDC and 37 ILC were labelled for NKX3.1. The IDC consisted of 20 luminal A, 7 luminal B, 14 Her2, and 45 triple negative carcinomas. The ILC consisted of 34 luminal A and 3 luminal B cases. NKX3.1 expression was scored as percentage nuclear labelling and labelling intensity. Nuclear NKX3.1 labelling was seen in 2 IDC (2%) and 10 ILCs (27%). labelling intensity was weak in all cases (1–100% nuclear positivity). Positive NKX3.1 labelling was significantly associated with ILC (p<0.0001). NKX3.1 labelling was seen only in ER and AR-positive carcinomas, which showed a significant correlation (p=0.0003 and p=0.0079, respectively). Expression was not correlated with tumour stage, size, Her2 expression, presence of lymph node metastases or age. This is the first study to evaluate NKX3.1 expression in breast carcinomas with known ER, PR, AR and Her2 status. Further studies are needed to evaluate what potential role NKX3.1 plays in ER and AR signalling and hormonal treatment response in breast carcinomas.
Sigma-Aldrich
NKX3.1 (EP356) Rabbit Monoclonal Primary Antibody