MilliporeSigma
  • CYP4F2 gene V433M polymorphism is associated with ischemic stroke in the male Northern Chinese Han population.
CYP4F2 gene V433M polymorphism is associated with ischemic stroke in the male Northern Chinese Han population.
Progress in neuro-psychopharmacology & biological psychiatry (2010-03-17)
Shumin Deng, Gang Zhu, Fang Liu, Hui Zhang, Xue Qin, Lei Li, He Zhiyi
ABSTRACT
CYP4F2 is a member of the cytochrome P450 enzymes and is responsible for metabolizing arachidonic acid to 20-hydroxyeicosatetraenoic acid (20-HETE); 20-HETE plays a role in the regulation of vascular tone in the cerebral, coronary, and renal circulation. The present study aimed to evaluate whether or not the CYP4F2 gene polymorphism V433M (rs2108622) is involved in ischemic stroke in the Northern Chinese Han population. In a case-control study, the participants included 302 (193 males and 109 females) patients with ischemic stroke and 350 (212 males and 138 females) healthy subjects. The V433M polymorphism of the CYP4F2 gene was analyzed by polymerase chain reaction (PCR) and restriction fragment length polymorphism (RFLP) with the restriction enzyme PvuII. The frequencies of the GG genotype and the G allele were higher in participants with ischemic stroke than in the control group (P=0.018). Multiple logistic regression analysis revealed the significance of the rs2108622 in males after adjustment for confounding factors. No difference was found in all participants and females. The GG genotype and G allele were associated with ischemic stroke in the male Northern Chinese Han population.