MilliporeSigma
  • Mutations in the gene phospholipase C, delta-1 (PLCD1) underlying hereditary leukonychia.
Mutations in the gene phospholipase C, delta-1 (PLCD1) underlying hereditary leukonychia.
European journal of dermatology : EJD (2012-11-15)
Hina Mir, Saadullah Khan, Muhammad Shoaib Arif, Ghazanfar Ali, Abdul Wali, Muhammad Ansar, Wasim Ahmad
ABSTRACT
Hereditary leukonychia or porcelain nails is a nail dystrophy characterized by whitening of the nail plates in all nails of the hands and feet. It may exhibit an autosomal recessive or autosomal dominant pattern of inheritance. Mutations in the gene PLCD1 have been reported to underlie hereditary leukonychia. In the present study, two Pakistani families with autosomal recessive hereditary leukonychia were investigated. Affected members of the families exhibited characteristic features of hereditary leukonychia with involvement of nails on both the hands and feet. Genotyping using microsatellite markers linked the families to the gene PLCD1 on chromosome 3p22.2. Sequence analysis of the gene detected one novel (p.Ser740ArgfsX19) and one previously reported mutation (p.Arg437X). This study expands spectrum of the mutations in the gene PLCD1 causing hereditary leukonychia.