MilliporeSigma
  • EphrinB-EphB receptor signaling contributes to neuropathic pain by regulating neural excitability and spinal synaptic plasticity in rats.
EphrinB-EphB receptor signaling contributes to neuropathic pain by regulating neural excitability and spinal synaptic plasticity in rats.
Pain (2008-05-02)
Xue-Jun Song, Ji-Hong Zheng, Jun-Li Cao, Wen-Tao Liu, Xue-Song Song, Zhi-Jiang Huang
ABSTRACT
Bidirectional signaling between ephrins and Eph receptor tyrosine kinases was first found to play important roles during development, but recently has been implicated in synaptic plasticity and pain processing in the matured nervous system. We show that ephrinB-EphB receptor signaling plays a critical role is induction and maintenance of neuropathic pain by regulating neural excitability and synaptic plasticity in the dorsal root ganglion (DRG) and the spinal dorsal horn (DH). Intrathecal application of blocking reagents for EphB-receptors, EphB1-Fc and EphB2-Fc chimeras inhibits the induction and maintenance of nerve injury-induced thermal hyperalgesia and mechanical allodynia. These blockers also prevent and suppress the nerve injury-induced hyperexcitability of nociceptive small DRG neurons, sensitization of DH neurons and long-term potentiation (LTP) of synapses between C fibers and DH neurons. In naïve, uninjured animals intrathecal administration of EphB-receptor activators ephrinB1-Fc and ephrinB2-Fc, respectively, induces thermal hypersensitivity and lowers the threshold for LTP, while EphB1-Fc prevents induction of the LTP. Western Blot analysis shows that nerve injury triggers an upregulation of the ephrinB1 and EphB1 receptor proteins in DRG and the spinal cord. These results indicate that, by regulating excitability of nociceptive-related neurons in DRG and DH and the synaptic plasticity at the spinal level, ephrinB-EphB receptor signaling contributes to neuropathic pain. This novel role for ephrinB-EphB receptor signaling suggests that these molecules may be useful therapeutic targets for treating pain after nerve injury.
Sigma-Aldrich
EphB2/Fc Chimera from mouse, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder