MilliporeSigma
Efficient RNA polyuridylation by noncanonical poly(A) polymerases.
Molecular and cellular biology (2007-03-14)
Olivia S Rissland, Andrea Mikulasova, Chris J Norbury
ABSTRACT
Nuclear poly(A) polymerase (PAP) polyadenylates nascent mRNAs, promoting their nuclear export, stability, and translation, while the related cytoplasmic polymerase GLD-2 activates translation of deadenylated mRNAs. Here we characterize the biochemical activity of fission yeast Schizosaccharomyces pombe Cid1, a putative cytoplasmic PAP implicated in cell cycle checkpoint controls. Surprisingly, Cid1 has robust poly(U) polymerase activity in vitro, especially when isolated in native multiprotein complexes. Furthermore, we found that upon S-phase arrest, the 3' ends of actin mRNAs were posttranscriptionally uridylated in a Cid1-dependent manner. Finally, Hs2 (ZCCHC6), a human ortholog of Cid1, shows similar activity. These data suggest that uridylation of mRNA forms the basis of an evolutionarily conserved mechanism of gene regulation.