MilliporeSigma
  • Efficient magnetic recycling of covalently attached enzymes on carbon-coated metallic nanomagnets.
Efficient magnetic recycling of covalently attached enzymes on carbon-coated metallic nanomagnets.
Bioconjugate chemistry (2014-03-29)
Vladimir Zlateski, Roland Fuhrer, Fabian M Koehler, Scott Wharry, Martin Zeltner, Wendelin J Stark, Thomas S Moody, Robert N Grass
ABSTRACT
In the pursuit of robust and reusable biocatalysts for industrial synthetic chemistry, nanobiotechnology is currently taking a significant part. Recently, enzymes have been immobilized on different nanoscaffold supports. Carbon coated metallic nanoparticles were found to be a practically useful support for enzyme immobilization due to their large surface area, high magnetic saturation, and manipulatable surface chemistry. In this study carbon coated cobalt nanoparticles were chemically functionalized (diazonium chemistry), activated for bioconjugation (N,N-disuccinimidyl carbonate), and subsequently used in enzyme immobilization. Three enzymes, β-glucosidase, α-chymotrypsin, and lipase B were successfully covalently immobilized on the magnetic nonsupport. The enzyme-particle conjugates formed retained their activity and stability after immobilization and were efficiently recycled from milliliter to liter scales in short recycle times.
Sigma-Aldrich
TurboBeads Silica, extent of labeling: SiO2 loading (Silica coating))
Sigma-Aldrich
TurboBeads Complexon, extent of labeling: ≥0.1 mmol/g loading (-Ph-CH2-N(CH2-COOH)2)