MilliporeSigma
  • Determination of aristolochic acid in botanicals and dietary supplements by liquid chromatography with ultraviolet detection and by liquid chromatography/mass spectrometry: single laboratory validation confirmation.
Determination of aristolochic acid in botanicals and dietary supplements by liquid chromatography with ultraviolet detection and by liquid chromatography/mass spectrometry: single laboratory validation confirmation.
Journal of AOAC International (2006-08-19)
William A Trujillo, Wendy R Sorenson, Paul La Luzerne, John W Austad, Darryl Sullivan
ABSTRACT
The presence of aristolochic acid in some dietary supplements is a concern to regulators and consumers. A method has been developed, by initially using a reference method as a guide, during single laboratory validation (SLV) for the determination of aristolochic acid I, also known as aristolochic acid A, in botanical species and dietary supplements at concentrations of approximately 2 to 32 microg/g. Higher levels were determined by dilution to fit the standard curve. Through the SLV, the method was optimized for quantification by liquid chromatography with ultraviolet detection (LC-UV) and LC/mass spectrometry (MS) confirmation. The test samples were extracted with organic solvent and water, then injected on a reverse phase LC column. Quantification was achieved with linear regression using a laboratory automation system. The SLV study included systematically optimizing the LC-UV method with regard to test sample size, fine grinding of solids, and solvent extraction efficiency. These parameters were varied in increments (and in separate optimization studies), in order to ensure that each parameter was individually studied; the test results include corresponding tables of parameter variations. In addition, the chromatographic conditions were optimized with respect to injection volume and detection wavelength. Precision studies produced overall relative standard deviation values from 2.44 up to 8.26% for aristolochic acid I. Mean recoveries were between 100 and 103% at the 2 microg/g level, between 102 and 103% at the 10 microg/g level, and 104% at the 30 microg/g level.