MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Actin-bundling protein TRIOBP forms resilient rootlets of hair cell stereocilia essential for hearing.
Actin-bundling protein TRIOBP forms resilient rootlets of hair cell stereocilia essential for hearing.
Cell (2010-06-01)
Shin-ichiro Kitajiri, Takeshi Sakamoto, Inna A Belyantseva, Richard J Goodyear, Ruben Stepanyan, Ikuko Fujiwara, Jonathan E Bird, Saima Riazuddin, Sheikh Riazuddin, Zubair M Ahmed, Jenny E Hinshaw, James Sellers, James R Bartles, John A Hammer, Guy P Richardson, Andrew J Griffith, Gregory I Frolenkov, Thomas B Friedman
ABSTRACT
Inner ear hair cells detect sound through deflection of mechanosensory stereocilia. Each stereocilium is supported by a paracrystalline array of parallel actin filaments that are packed more densely at the base, forming a rootlet extending into the cell body. The function of rootlets and the molecules responsible for their formation are unknown. We found that TRIOBP, a cytoskeleton-associated protein mutated in human hereditary deafness DFNB28, is localized to rootlets. In vitro, purified TRIOBP isoform 4 protein organizes actin filaments into uniquely dense bundles reminiscent of rootlets but distinct from bundles formed by espin, an actin crosslinker in stereocilia. We generated mutant Triobp mice (Triobp(Deltaex8/Deltaex8)) that are profoundly deaf. Stereocilia of Triobp(Deltaex8/Deltaex8) mice develop normally but fail to form rootlets and are easier to deflect and damage. Thus, F-actin bundling by TRIOBP provides durability and rigidity for normal mechanosensitivity of stereocilia and may contribute to resilient cytoskeletal structures elsewhere.