MilliporeSigma
  • Nucleophilic β-Carbon Activation of Propionic Acid as a 3-Carbon Synthon by Carbene Organocatalysis.
Nucleophilic β-Carbon Activation of Propionic Acid as a 3-Carbon Synthon by Carbene Organocatalysis.
Chemistry (Weinheim an der Bergstrasse, Germany) (2015-05-28)
Zhichao Jin, Ke Jiang, Zhenqian Fu, Jaume Torres, Pengcheng Zheng, Song Yang, Bao-An Song, Yonggui Robin Chi
ABSTRACT
Direct β-carbon activation of propionic acid (C2H5CO2H) by carbene organocatalysis has been developed. This activation affords the smallest azolium homoenolate intermediate (without any substituent) as a 3-carbon nucleophile for enantioselective reactions. Propionic acid is an excellent raw material because it is cheap, stable, and safe. This approach provides a much better solution to azolium homoenolate synthesis than the previously established use of acrolein (enal without any substituent), which is expensive, unstable, and toxic.
Sigma-Aldrich
Propionic acid, puriss. p.a., ≥99.5% (GC)