MilliporeSigma
  • The effect of temperature on supported dipalmitoylphosphatidylcholine (DPPC) bilayers: structure and lubrication performance.
The effect of temperature on supported dipalmitoylphosphatidylcholine (DPPC) bilayers: structure and lubrication performance.
Journal of colloid and interface science (2015-01-18)
Min Wang, Thomas Zander, Xiaoyan Liu, Chao Liu, Akanksha Raj, D C Florian Wieland, Vasil M Garamus, Regine Willumeit-Römer, Per Martin Claesson, Andra Dėdinaitė
ABSTRACT
Phospholipids fulfill an important role in joint lubrication. They, together with hyaluronan and glycoproteins, are the biolubricants that sustain low friction between cartilage surfaces bathed in synovial fluid. In this work we have investigated how the friction force and load bearing capacity of 1,2-dipalmitoyl-sn-glycero-3-phosphocholine (DPPC) bilayers on silica surfaces are affected by temperature, covering the temperature range 25-52°C. Friction forces have been determined utilizing the AFM colloidal probe technique, which showed that DPPC bilayers are able to provide low friction forces over the whole temperature interval. However, the load bearing capacity is improved at higher temperatures. We interpret this finding as being a consequence of lower rigidity and higher self-healing capacity of the DPPC bilayer in the liquid disordered state compared to the gel state. The corresponding structure of solid supported DPPC bilayers at the silica-liquid interface has been followed using X-ray reflectivity measurements, which suggests that the DPPC bilayer is in the gel phase at 25°C and 39°C and in the liquid disordered state at 55°C. Well-defined bilayer structures were observed for both phases. The deposited DPPC bilayers were also imaged using AFM PeakForce Tapping mode, and these measurements indicated a less homogeneous layer at temperatures below 37°C.
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand
Sigma-Aldrich
Phosphate buffered saline, tablet