MilliporeSigma
  • Gas chromatographic determination of organomercury following aqueous derivatization with sodium tetraethylborate and sodium tetraphenylborate. Comparative study of gas chromatography coupled with atomic fluorescence spectrometry, atomic emission spectrometry and mass spectrometry.
Gas chromatographic determination of organomercury following aqueous derivatization with sodium tetraethylborate and sodium tetraphenylborate. Comparative study of gas chromatography coupled with atomic fluorescence spectrometry, atomic emission spectrometry and mass spectrometry.
Journal of chromatography. A (2000-05-24)
Y Cai, S Monsalud, R Jaffé, R D Jones
ABSTRACT
Several hyphenated analytical techniques, including gas chromatography (GC) coupled with atomic fluorescence spectrometry (AFS), microwave-induced plasma atomic emission spectrometry (AES), and mass spectrometry (MS), have been evaluated for methylmercury and ethylmercury analysis following aqueous derivatization with both sodium tetraethylborate and sodium tetraphenylborate. Both GC-AFS and GC-AES were shown to be excellent techniques with detection limits in the range of sub-picogram levels (0.02-0.04 pg as Hg). Both techniques have wide linear ranges, although setting of the AFS sensitivity has to be selected manually based on the concentration of mercury in the sample. Phenylation seems to be more favorable in this study because of its capability of distinguishing between ethylmercury and inorganic mercury, and low cost compared to ethylation. Although sensitivity of GC-MS is poor with detection limits ranging from 30 to 50 pg as Hg, it is an essential technique for confirmation of the derivatization products.
Sigma-Aldrich
Sodium tetraphenylborate, puriss. p.a., ACS reagent, ≥99.5% (NT)