MilliporeSigma
  • Telomerase-based pharmacologic enhancement of antiviral function of human CD8+ T lymphocytes.
Telomerase-based pharmacologic enhancement of antiviral function of human CD8+ T lymphocytes.
Journal of immunology (Baltimore, Md. : 1950) (2008-11-05)
Steven Russell Fauce, Beth D Jamieson, Allison C Chin, Ronald T Mitsuyasu, Stan T Parish, Hwee L Ng, Christina M Ramirez Kitchen, Otto O Yang, Calvin B Harley, Rita B Effros
ABSTRACT
Telomerase reverse transcribes telomere DNA onto the ends of linear chromosomes and retards cellular aging. In contrast to most normal somatic cells, which show little or no telomerase activity, immune cells up-regulate telomerase in concert with activation. Nevertheless, during aging and chronic HIV-1 infection, there are high proportions of dysfunctional CD8(+) CTL with short telomeres, suggesting that telomerase is limiting. The present study shows that exposure of CD8(+) T lymphocytes from HIV-infected human donors to a small molecule telomerase activator (TAT2) modestly retards telomere shortening, increases proliferative potential, and, importantly, enhances cytokine/chemokine production and antiviral activity. The enhanced antiviral effects were abrogated in the presence of a potent and specific telomerase inhibitor, suggesting that TAT2 acts primarily through telomerase activation. Our study is the first to use a pharmacological telomerase-based approach to enhance immune function, thus directly addressing the telomere loss immunopathologic facet of chronic viral infection.
Sigma-Aldrich
PD 98059, PD 98059, CAS 167869-21-8, is a cell-permeable, selective & reversible inhibitor of MAP Kinase Kinase (MEK). Inhibits MAP Kinase activation and subsequent phosphorylation of MAP Kinase substrates.
Sigma-Aldrich
TRAPeze RT Telomerase Detection Kit, A highly sensitive in vitro assay for the fluorometric detection & real time quantification of telomerase activity in cells.
Sigma-Aldrich
Akt Inhibitor III, The Akt Inhibitor III controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Akt Inhibitor, The Akt Inhibitor controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Cycloastragenol, ≥98% (HPLC)