MilliporeSigma
  • Leukemia-associated Rho guanine nucleotide exchange factor (LARG) links heterotrimeric G proteins of the G(12) family to Rho.
Leukemia-associated Rho guanine nucleotide exchange factor (LARG) links heterotrimeric G proteins of the G(12) family to Rho.
FEBS letters (2000-11-30)
S Fukuhara, H Chikumi, J S Gutkind
ABSTRACT
A putative guanine nucleotide exchange factor (GEF), termed leukemia-associated RhoGEF (LARG), was recently identified upon fusion to the coding sequence of the MLL gene in acute myeloid leukemia. Although the function of LARG is still unknown, it exhibits a number of structural domains suggestive of a role in signal transduction, including a PDZ domain, a LH/RGS domain, and a Dbl homology/pleckstrin homology domain. Here, we show that LARG can activate Rho in vivo. Furthermore, we present evidence that LARG is an integral component of a novel biochemical route whereby G protein-coupled receptors (GPCRs) and heterotrimeric G proteins of the G alpha(12) family stimulate Rho-dependent signaling pathways.