MilliporeSigma
  • Resuscitation by ferrioxamine E of stressed Salmonella enterica serovar typhimurium from soil and water microcosms.
Resuscitation by ferrioxamine E of stressed Salmonella enterica serovar typhimurium from soil and water microcosms.
Applied and environmental microbiology (2000-08-31)
R Reissbrodt, H Heier, H Tschäpe, R A Kingsley, P H Williams
ABSTRACT
Storage of Salmonella enterica serovar Typhimurium strains in soil and water microcosms resulted in loss of culturability on standard plating media. Prior incubation in buffered peptone water supplemented with ferrioxamine E markedly extended the time that bacteria were recoverable by plating, except in the case of mutants deficient in ferrioxamine E uptake.
Product Description
Millipore
Ferrioxamine E from Streptomyces antibioticus, suitable for microbiology, ≥95%