MilliporeSigma
  • Enigma prevents Cbl-c-mediated ubiquitination and degradation of RETMEN2A.
Enigma prevents Cbl-c-mediated ubiquitination and degradation of RETMEN2A.
PloS one (2014-01-28)
Stephen C Kales, Marion M Nau, Anand S Merchant, Stanley Lipkowitz
ABSTRACT
The Cbl proteins (Cbl, Cbl-b, and Cbl-c) are a highly conserved family of RING finger ubiquitin ligases (E3s) that function as negative regulators of tyrosine kinases in a wide variety of signal transduction pathways. In this study, we identify a new Cbl-c interacting protein, Enigma (PDLIM7). This interaction is specific to Cbl-c as Enigma fails to bind either of its closely related homologues, Cbl and Cbl-b. The binding between Enigma and Cbl-c is mediated through the LIM domains of Enigma as removal of all three LIM domains abrogates this interaction, while only LIM1 is sufficient for binding. Here we show that Cbl-c binds wild-type and MEN2A isoforms of the receptor tyrosine kinase, RET, and that Cbl-c enhances ubiquitination and degradation of activated RET. Enigma blocks Cbl-c-mediated RETMEN2A ubiquitination and degradation. Cbl-c decreased downstream ERK activation by RETMEN2A and co-expression of Enigma blocked the Cbl-c-mediated decrease in ERK activation. Enigma showed no detectable effect on Cbl-c-mediated ubiquitination of activated EGFR suggesting that this effect is specific to RET. Through mapping studies, we show that Cbl-c and Enigma bind RETMEN2A at different residues. However, binding of Enigma to RETMENA prevents Cbl-c recruitment to RETMEN2A. Consistent with these biochemical data, exploratory analyses of breast cancer patients with high expression of RET suggest that high expression of Cbl-c correlates with a good outcome, and high expression of Enigma correlates with a poor outcome. Together, these data demonstrate that Cbl-c can ubiquitinate and downregulate RETMEN2A and implicate Enigma as a positive regulator of RETMEN2A through blocking of Cbl-mediated ubiquitination and degradation.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
MG-132, A cell-permeable, potent, reversible proteasome inhibitor (Ki = 4 nM).
Roche
Anti-HA Affinity Matrix, from rat IgG1