MilliporeSigma
  • The transmembrane adapter protein SIT regulates thymic development and peripheral T-cell functions.
The transmembrane adapter protein SIT regulates thymic development and peripheral T-cell functions.
Molecular and cellular biology (2005-08-19)
Luca Simeoni, Vilmos Posevitz, Uwe Kölsch, Ines Meinert, Eddy Bruyns, Klaus Pfeffer, Dirk Reinhold, Burkhart Schraven
ABSTRACT
SIT is a transmembrane adapter protein that modulates signals emanating from the T-cell receptor (TCR). Here, we have used gene-targeted mice to assess the role of SIT for T-cell development and peripheral T-cell functions. SIT(-/-) double-positive thymocytes show an upregulation of the activation markers CD5 and CD69, suggesting that SIT negatively regulates TCR-mediated signals at the CD4(+) CD8(+) stage of thymic development. This assumption is further supported by the observation that in female H-Y TCR transgenic mice, positive selection is enhanced and even converted to negative selection. Similarly, mature peripheral T cells are hyperresponsive towards TCR-mediated stimuli and produce larger amounts of T-helper 1 (TH1) cytokines, and SIT-deficient mice show an increased susceptibility to develop experimental autoimmune encephalomyelitis, a mouse model of multiple sclerosis. These results demonstrate that SIT is a critical negative regulator of TCR-mediated signaling and finely tunes the signals required for thymic selection and peripheral T-cell activation.