MilliporeSigma
Efficacy of anti-IL-1 treatment in Majeed syndrome.
Annals of the rheumatic diseases (2012-10-23)
Troels Herlin, Bente Fiirgaard, Mette Bjerre, Gitte Kerndrup, Henrik Hasle, Xinyu Bing, Polly J Ferguson
ABSTRACT
Majeed syndrome is an autosomal recessive disorder characterised by the triad of chronic recurrent multifocal osteomyelitis, congenital dyserythropoietic anaemia and a neutrophilic dermatosis that is caused by mutations in LPIN2. Long-term outcome is poor. This is the first report detailing the treatment of Majeed syndrome with biological agents and demonstrates clinical improvement with IL-1blockade. We describe the clinical presentation, genetic analysis, cytokine profiles and response to biological therapy in two brothers with Majeed syndrome. Both boys were homozygous for a novel 2-base pair deletion in LPIN2 (c.1312_1313delCT; p.Leu438fs+16X), confirming the diagnosis. Their bone disease and anaemia were refractory to treatment with corticosteroids. Both siblings had elevated proinflammatory cytokines in their serum, including tumour necrosis factor α (TNF-α), however a trial of the TNF inhibitor etanercept resulted in no improvement. IL-1 inhibition with either a recombinant IL-1 receptor antagonist (anakinra) or an anti-IL-1β antibody (canakinumab) resulted in dramatic clinical and laboratory improvement. The differential response to treatment with TNF-α or IL-1 blocking agents sheds light into disease pathogenesis; it supports the hypothesis that Majeed syndrome is an IL-1β dependent autoinflammatory disorder, and further underscores the importance of IL-1 in sterile bone inflammation.