MilliporeSigma
  • Aciculin interacts with filamin C and Xin and is essential for myofibril assembly, remodeling and maintenance.
Aciculin interacts with filamin C and Xin and is essential for myofibril assembly, remodeling and maintenance.
Journal of cell science (2014-06-26)
Sibylle Molt, John B Bührdel, Sergiy Yakovlev, Peter Schein, Zacharias Orfanos, Gregor Kirfel, Lilli Winter, Gerhard Wiche, Peter F M van der Ven, Wolfgang Rottbauer, Steffen Just, Alexey M Belkin, Dieter O Fürst
ABSTRACT
Filamin C (FLNc) and Xin actin-binding repeat-containing proteins (XIRPs) are multi-adaptor proteins that are mainly expressed in cardiac and skeletal muscles and which play important roles in the assembly and repair of myofibrils and their attachment to the membrane. We identified the dystrophin-binding protein aciculin (also known as phosphoglucomutase-like protein 5, PGM5) as a new interaction partner of FLNc and Xin. All three proteins colocalized at intercalated discs of cardiac muscle and myotendinous junctions of skeletal muscle, whereas FLNc and aciculin also colocalized in mature Z-discs. Bimolecular fluorescence complementation experiments in developing cultured mammalian skeletal muscle cells demonstrated that Xin and aciculin also interact in FLNc-containing immature myofibrils and areas of myofibrillar remodeling and repair induced by electrical pulse stimulation (EPS). Fluorescence recovery after photobleaching (FRAP) experiments showed that aciculin is a highly dynamic and mobile protein. Aciculin knockdown in myotubes led to failure in myofibril assembly, alignment and membrane attachment, and a massive reduction in myofibril number. A highly similar phenotype was found upon depletion of aciculin in zebrafish embryos. Our results point to a thus far unappreciated, but essential, function of aciculin in myofibril formation, maintenance and remodeling.