MilliporeSigma
MAGOH interacts with a novel RNA-binding protein.
Genomics (2000-02-09)
X F Zhao, N J Nowak, T B Shows, P D Aplan
ABSTRACT
MAGOH is the human homologue of Drosophila mago nashi, a protein that is required for normal germ plasm development in the Drosophila embryo. Using human MAGOH as a bait protein in a yeast two-hybrid screen, we recovered four independent cDNA clones that encode different lengths of a novel protein containing a conserved RNA-binding region. This gene, designated RBM8, encodes a 173-aa protein that was shown to have an apparent molecular mass of 26 kDa, as demonstrated by in vitro translation assay. The interaction between MAGOH and RBM8 was demonstrated by both yeast two-hybrid and GST fusion protein pull-down assays. Like MAGOH, RBM8 gene is expressed ubiquitously in human tissues; three species of RBM8 mRNA were detected. Also similar to MAGOH, RBM8 expression is serum inducible in quiescent NIH3T3 fibroblast cells.