MilliporeSigma
  • Human PIH1 associates with histone H4 to mediate the glucose-dependent enhancement of pre-rRNA synthesis.
Human PIH1 associates with histone H4 to mediate the glucose-dependent enhancement of pre-rRNA synthesis.
Journal of molecular cell biology (2012-03-01)
Niu Zhai, Zhong-liang Zhao, Mo-bin Cheng, Yu-wei Di, Hai-xia Yan, Chun-yu Cao, Hui Dai, Ye Zhang, Yu-fei Shen
ABSTRACT
Ribosome biogenesis is critical in the growth of eukaryotic cells, in which the synthesis of precursor ribosomal RNA is the first and rate-limiting step. Here, we show that human PIH1 domain-containing protein 1 (PIH1) interacts directly with histone H4 and recruits the Brg1-SWI/SNF complex via SNF5 to human rRNA genes. This process is likely involved in PIH1-dependent DNase I-hypersensitive chromatin remodeling at the core promoter of the rRNA genes. PIH1 mediates the occupancy of not only the Brg1 complex but also the Pol I complex at the core promoter and enhances transcription initiation of rRNA genes. Additionally, the interaction between PIH1 and H4K16 expels TIP5, a component of the silencing nucleolar remodeling complex (NoRC), from the core region, suggesting that PIH1 is involved in the derepression of NoRC-silenced rRNA genes. These data indicate that PIH1 is a positive regulator of human rRNA genes and is of great importance for the recovery of human cells from nutrient starvation and the transition to glucose-induced exponential growth in vivo.