MilliporeSigma
  • Genome scans and gene expression microarrays converge to identify gene regulatory loci relevant in schizophrenia.
Genome scans and gene expression microarrays converge to identify gene regulatory loci relevant in schizophrenia.
Human genetics (2006-04-11)
Marquis P Vawter, Mary E Atz, Brandi L Rollins, Kathleen M Cooper-Casey, Ling Shao, William F Byerley
ABSTRACT
Multiple linkage regions have been reported in schizophrenia, and some appear to harbor susceptibility genes that are differentially expressed in postmortem brain tissue derived from unrelated individuals. We combined traditional genome-wide linkage analysis in a multiplex family with lymphocytic genome-wide expression analysis. A genome scan suggested linkage to a chromosome 4q marker (D4S1530, LOD 2.17, theta = 0) using a dominant model. Haplotype analysis using flanking microsatellite markers delineated a 14 Mb region that cosegregated with all those affected. Subsequent genome-wide scan with SNP genotypes supported the evidence of linkage to 4q33-35.1 (LOD = 2.39) using a dominant model. Genome-wide microarray analysis of five affected and five unaffected family members identified two differentially expressed genes within the haplotype AGA and GALNT7 (aspartylglucosaminidase and UDP-N-acetyl-alpha-D-galactosamine: polypeptide N-acetylgalactosaminyltransferase 7) with nominal significance; however, these genes did not remain significant following analysis of covariance. We carried out genome-wide linkage analyses between the quantitative expression phenotype and genetic markers. AGA expression levels showed suggestive linkage to multiple markers in the haplotype (maximum LOD = 2.37) but to no other genomic region. GALNT7 expression levels showed linkage to regulatory loci at 4q28.1 (maximum LOD = 3.15) and in the haplotype region at 4q33-35.1 (maximum LOD = 2.37). ADH1B (alcohol dehydrogenase IB) was linked to loci at 4q21-q23 (maximum LOD = 3.08) and haplotype region at 4q33-35.1 (maximum LOD = 2.27). Seven differentially expressed genes were validated with RT-PCR. Three genes in the 4q33-35.1 haplotype region were also differentially expressed in schizophrenia in postmortem dorsolateral prefrontal cortex: AGA, HMGB2, and SCRG1. These results indicate that combining differential gene expression with linkage analysis may help in identifying candidate genes and potential regulatory sites. Moreover, they also replicate recent findings of complex trans- and cis- regulation of genes.