- A practical approach to crosslinking.
A practical approach to crosslinking.
Molecular biology reports (1993-04-01)
G Mattson, E Conklin, S Desai, G Nielander, M D Savage, S Morgensen
PMID8326953
ABSTRACT
The various aspects of chemical crosslinking are addressed. Crosslinker reactivity, specificity, spacer arm length and solubility characteristics are detailed. Considerations for choosing one of these crosslinkers for a particular application are given as well as reaction conditions and practical tips for use of each category of crosslinkers.