MilliporeSigma
  • Functional platelet defects in children with severe chronic ITP as tested with 2 novel assays applicable for low platelet counts.
Functional platelet defects in children with severe chronic ITP as tested with 2 novel assays applicable for low platelet counts.
Blood (2014-01-05)
Esther R van Bladel, Annemieke G Laarhoven, Laila B van der Heijden, Katja M Heitink-Pollé, Leendert Porcelijn, C Ellen van der Schoot, Masja de Haas, Mark Roest, Gestur Vidarsson, Philip G de Groot, Marrie C A Bruin
ABSTRACT
Immune thrombocytopenia (ITP) is an autoimmune disease with a complex heterogeneous pathogenesis and a bleeding phenotype that is not necessarily correlated to platelet count. In this study, the platelet function was assessed in a well-defined cohort of 33 pediatric chronic ITP patients. Because regular platelet function test cannot be performed in patients with low platelet counts, 2 new assays were developed to determine platelet function: first, the microaggregation test, measuring in platelets isolated from 10 mL of whole blood the platelet potential to form microaggregates in response to an agonist; second, the platelet reactivity assay, measuring platelet reactivity to adenosine diphosphate (ADP), convulxin (CVX), and thrombin receptor activator peptide in only 150 μL of unprocessed whole blood. Patients with a severe bleeding phenotype demonstrated a decreased aggregation potential upon phorbol myristate acetate stimulation, decreased platelet degranulation following ADP stimulation, and a higher concentration of ADP and CVX needed to activate the glycoprotein IIbIIIa complex compared with patients with a mild bleeding phenotype. In conclusion, here we have established 2 functional tests that allow for evaluation of platelet function in patients with extremely low platelet counts (<10(9)). These tests show that platelet function is related to bleeding phenotype in chronic ITP.
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
PKH26 Red Fluorescent Cell Linker Midi Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies