MilliporeSigma
  • A pharmacological map of the PI3-K family defines a role for p110alpha in insulin signaling.
A pharmacological map of the PI3-K family defines a role for p110alpha in insulin signaling.
Cell (2006-05-02)
Zachary A Knight, Beatriz Gonzalez, Morri E Feldman, Eli R Zunder, David D Goldenberg, Olusegun Williams, Robbie Loewith, David Stokoe, Andras Balla, Balazs Toth, Tamas Balla, William A Weiss, Roger L Williams, Kevan M Shokat
ABSTRACT
Phosphoinositide 3-kinases (PI3-Ks) are an important emerging class of drug targets, but the unique roles of PI3-K isoforms remain poorly defined. We describe here an approach to pharmacologically interrogate the PI3-K family. A chemically diverse panel of PI3-K inhibitors was synthesized, and their target selectivity was biochemically enumerated, revealing cryptic homologies across targets and chemotypes. Crystal structures of three inhibitors bound to p110gamma identify a conformationally mobile region that is uniquely exploited by selective compounds. This chemical array was then used to define the PI3-K isoforms required for insulin signaling. We find that p110alpha is the primary insulin-responsive PI3-K in cultured cells, whereas p110beta is dispensable but sets a phenotypic threshold for p110alpha activity. Compounds targeting p110alpha block the acute effects of insulin treatment in vivo, whereas a p110beta inhibitor has no effect. These results illustrate systematic target validation using a matrix of inhibitors that span a protein family.
Sigma-Aldrich
PI 3-Kα Inhibitor VIII, The PI 3-Kα Inhibitor VIII controls the biological activity of PI 3-Kα. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
PI-103, A cell-permeable pyridinylfuranopyrimidine compound that acts as a potent and ATP-competitive inhibitor of DNA-PK, PI3-K, and mTOR.