MilliporeSigma
Chemical constituents of Allophylus longipes.
Chinese journal of natural medicines (2013-01-11)
Xiang-Yun Zhang, Xiang-Hai Cai, Xiao-Dong Luo
ABSTRACT
To investigate the chemical constituents of Allophylus longipes. Compounds were isolated and purified by various chromatographic techniques and their structures were elucidated by physicochemical characteristics and spectral data. Twenty-five compounds were isolated and identified as cycloart-24-en-3β, 26-diol (1), 3-oxotrirucalla-7, 24-dien-21-oic acid (2), zizyberenalic acid (3), colubrinic acid (4), ent-4(15)-eudesmene-1β, 6α-diol (5), 4(15)-eudesmene-1β, 8α-diol (6), 4(15)-eudesmene-1β, 5α-diol (7), methyl asterrate (8), betulin (9), betulinic aldehyde (10), betulinic acid (11), 3β-hydroxy-5α, 8α-epidioxyergosta-6, 22-dien (12), 3-oxo-19α-hydroxyurs-12-en-28-oic acid (13), ursolic acid (14), scopoletin (15), fraxidin (16), cleomiscosin A (17), 4-hydroxy-3-methoxybenzaldehyde (18), 4-hydroxy-3-methoxycinnamaldehyde (19), 2',6'-dihydroxy-4'-methoxyacetophenone (20), p-(aminoalkyl)-benzoic acid (21), 4-hydroxy-3-methoxybenzoic acid (22), 1-O-p-coumaroylglucose (23), β-sitosterol (24), and poriferast-5-ene-3β, 4β-diol (25). All the compounds were isolated from Allophylus longipes for the first time.
Sigma-Aldrich
Methyl asterrate, ≥95% (LC/MS-UV)