MilliporeSigma
  • Methylphenidate treatment beyond adolescence maintains increased cocaine self-administration in the spontaneously hypertensive rat model of attention deficit/hyperactivity disorder.
Methylphenidate treatment beyond adolescence maintains increased cocaine self-administration in the spontaneously hypertensive rat model of attention deficit/hyperactivity disorder.
Pharmacology, biochemistry, and behavior (2015-02-04)
Britahny M Baskin, Linda P Dwoskin, Kathleen M Kantak
ABSTRACT
Past research with the spontaneously hypertensive rat (SHR) model of attention deficit/hyperactivity disorder showed that adolescent methylphenidate treatment enhanced cocaine abuse risk in SHR during adulthood. The acquisition of cocaine self-administration was faster, and cocaine dose-response functions were shifted upward under fixed-ratio and progressive ratio schedules compared to adult SHR that received adolescent vehicle treatment or to control strains that received adolescent methylphenidate treatment. The current study determined if extending treatment beyond adolescence would ameliorate long-term consequences of adolescent methylphenidate treatment on cocaine abuse risk in adult SHR. Treatments (vehicle or 1.5mg/kg/day oral methylphenidate) began on postnatal day 28. Groups of male SHR were treated with vehicle during adolescence and adulthood, with methylphenidate during adolescence and vehicle during adulthood, or with methylphenidate during adolescence and adulthood. The group receiving adolescent-only methylphenidate was switched to vehicle on P56. Cocaine self-administration began on postnatal day 77, and groups receiving methylphenidate during adolescence and adulthood were treated either 1-h before or 1-h after daily sessions. At baseline under a fixed-ratio 1 schedule, cocaine self-administration (2h sessions; 0.3mg/kg unit dose) did not differ among the four treatment groups. Under a progressive ratio schedule (4.5h maximum session length; 0.01-1.0mg/kg unit doses), breakpoints for self-administered cocaine in SHR receiving the adult methylphenidate treatment 1-h pre-session were not different from the vehicle control group. However, compared to the vehicle control group, breakpoints for self-administered cocaine at the 0.3 and 1.0mg/kg unit doses were greater in adult SHR that received adolescent-only methylphenidate or received methylphenidate that was continued into adulthood and administered 1-h post-session. These findings suggest that extending methylphenidate treatment beyond adolescence does not ameliorate explicitly the long-term consequences of adolescent methylphenidate treatment. Pre-session methylphenidate may mask temporarily the detection of an increase in cocaine self-administration following chronic methylphenidate treatment.