MilliporeSigma
  • Breast-feeding improves gut maturation compared with formula feeding in preterm babies.
Breast-feeding improves gut maturation compared with formula feeding in preterm babies.
Journal of pediatric gastroenterology and nutrition (2014-08-12)
Kostan W Reisinger, Loes de Vaan, Boris W Kramer, Tim G A M Wolfs, L W Ernest van Heurn, Joep P M Derikx
ABSTRACT
The incidence of necrotizing enterocolitis (NEC) is higher in formula-fed babies than in breast-fed babies, which may be caused by breast-feeding-induced gut maturation. The effect of breast-feeding on gut maturation has been widely studied in animal models. This study aimed to assess the effects of breast-feeding on intestinal maturation in prematurely born babies by evaluating postnatal changes in urinary intestinal fatty acid binding protein (I-FABP) levels, a specific enterocyte marker. Gut maturation in 40 premature babies (<37 weeks of gestation) without gastrointestinal morbidity was studied, of whom 21 were exclusively breast-fed and 19 were formula-fed infants. Urinary I-FABP levels as the measure of gut maturation were measured at 5, 12, 19, and 26 days after birth. In breast-fed infants, there was a significant increase in median urinary I-FABP levels between 5 and 12 days after birth (104 [78-340] pg/mL to 408 [173-1028] pg/mL, P = 0.002), whereas I-FABP concentration in formula-fed infants increased between 12 and 19 days after birth (105 [44-557] pg/mL, 723 [103-1670] pg/mL, P = 0.004). Breast-fed babies had significantly higher median urinary I-FABP levels at postnatal day 12 (P = 0.01). The time course of the postnatal increase in urinary I-FABP levels reflecting gut maturation was significantly delayed in formula-fed babies, suggesting a delayed physiological response in formula-fed compared with breast-fed infants.
Sigma-Aldrich
Lactulose, ≥98.0% (HPLC)
Sigma-Aldrich
Lactulose, ≥95% (HPLC)
Lactulose, European Pharmacopoeia (EP) Reference Standard
USP
Lactulose, United States Pharmacopeia (USP) Reference Standard
Supelco
Lactulose, Pharmaceutical Secondary Standard; Certified Reference Material