MilliporeSigma
  • Genome sequence of mungbean and insights into evolution within Vigna species.
Genome sequence of mungbean and insights into evolution within Vigna species.
Nature communications (2014-11-12)
Yang Jae Kang, Sue K Kim, Moon Young Kim, Puji Lestari, Kil Hyun Kim, Bo-Keun Ha, Tae Hwan Jun, Won Joo Hwang, Taeyoung Lee, Jayern Lee, Sangrea Shim, Min Young Yoon, Young Eun Jang, Kwang Soo Han, Puntaree Taeprayoon, Na Yoon, Prakit Somta, Patcharin Tanya, Kwang Soo Kim, Jae-Gyun Gwag, Jung-Kyung Moon, Yeong-Ho Lee, Beom-Seok Park, Aureliano Bombarely, Jeffrey J Doyle, Scott A Jackson, Roland Schafleitner, Peerasak Srinives, Rajeev K Varshney, Suk-Ha Lee
ABSTRACT
Mungbean (Vigna radiata) is a fast-growing, warm-season legume crop that is primarily cultivated in developing countries of Asia. Here we construct a draft genome sequence of mungbean to facilitate genome research into the subgenus Ceratotropis, which includes several important dietary legumes in Asia, and to enable a better understanding of the evolution of leguminous species. Based on the de novo assembly of additional wild mungbean species, the divergence of what was eventually domesticated and the sampled wild mungbean species appears to have predated domestication. Moreover, the de novo assembly of a tetraploid Vigna species (V. reflexo-pilosa var. glabra) provides genomic evidence of a recent allopolyploid event. The species tree is constructed using de novo RNA-seq assemblies of 22 accessions of 18 Vigna species and protein sets of Glycine max. The present assembly of V. radiata var. radiata will facilitate genome research and accelerate molecular breeding of the subgenus Ceratotropis.
Sigma-Aldrich
NSC 632839, ≥98% (HPLC)