MilliporeSigma
  • Knockdown of response gene to complement 32 (RGC32) induces apoptosis and inhibits cell growth, migration, and invasion in human lung cancer cells.
Knockdown of response gene to complement 32 (RGC32) induces apoptosis and inhibits cell growth, migration, and invasion in human lung cancer cells.
Molecular and cellular biochemistry (2014-05-17)
Ran Xu, Chao Shang, Jungang Zhao, Yun Han, Jun Liu, Kuanbing Chen, Wenjun Shi
ABSTRACT
Response gene to complement 32 (RGC32) is a novel protein originally identified as a cell cycle activator and has been demonstrated to be overexpressed in a variety of human malignancies, including lung cancer. However, the potential role of RGC32 in lung cancer initiation and progression remains to be elucidated. In the present study, RNA interference mediated by plasmid expressing RGC32 short-hairpin RNA (shRNA) was utilized to knockdown RGC32 expression in human lung cancer LTE cells. We found that the mRNA and protein expression levels of RGC32 were significantly decreased in RGC32-specific shRNA-transfected cells in comparison with the untransfected and control shRNA-transfected cells. Furthermore, knockdown of RGC32 dramatically reduced cell proliferation, colony formation, and invasion and migration capacities of LTE cells in vitro. Specific down-regulation of RGC32 caused G0/G1 cell cycle arrest and eventual apoptosis. Meanwhile, Western blot analysis indicated that cells with stably knockdown of RGC32 showed decreased expression levels of Cyclin D1, Cyclin E, Bcl-2, matrix metalloproteinase (MMP)-2, and MMP-9, but increased expression levels of activate caspase-3, Bax, and cleaved poly (ADP-ribose) polymerase (PARP) in comparison with control shRNA-transfected cells. Taken together, our data suggest that RGC32 is involved in tumorigenesis of human lung cancer and may serve as a promising therapeutic target for lung cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse 1190002h23rik