MilliporeSigma
  • Fatty acid-binding protein E-FABP restricts tumor growth by promoting IFN-β responses in tumor-associated macrophages.
Fatty acid-binding protein E-FABP restricts tumor growth by promoting IFN-β responses in tumor-associated macrophages.
Cancer research (2014-04-10)
Yuwen Zhang, Yanwen Sun, Enyu Rao, Fei Yan, Qiang Li, Ying Zhang, Kevin A T Silverstein, Shujun Liu, Edward Sauter, Margot P Cleary, Bing Li
ABSTRACT
Fatty acid-binding proteins (FABP) are known central regulators of both metabolic and inflammatory pathways, but their role in tumor development remains largely unexplored. Here, we report that host expression of epidermal FABP (E-FABP) protects against mammary tumor growth. We find that E-FABP is highly expressed in macrophages, particularly in a specific subset, promoting their antitumor activity. In the tumor stroma, E-FABP-expressing tumor-associated macrophages (TAM) produce high levels of IFN-β through upregulation of lipid droplet formation in response to tumors. E-FABP-mediated IFN-β signaling can further enhance recruitment of tumoricidal effector cells, in particular natural killer cells, to the tumor stroma for antitumor activity. These findings identify E-FABP as a new protective factor to strengthen IFN-β responses against tumor growth.
Sigma-Aldrich
Human IFNA1 / Interferon Alpha-1/13 ELISA Kit, for serum, plasma, cell culture supernatants and urine
Sigma-Aldrich
Human IFN γ ELISA Kit, for cell and tissue lysates
Sigma-Aldrich
Human IFN γ ELISA Kit, for serum, plasma, cell culture supernatant and urine
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fabp5