MilliporeSigma
  • The herbicide glyphosate causes behavioral changes and alterations in dopaminergic markers in male Sprague-Dawley rat.
The herbicide glyphosate causes behavioral changes and alterations in dopaminergic markers in male Sprague-Dawley rat.
Neurotoxicology (2014-12-20)
Isela Hernández-Plata, Magda Giordano, Mauricio Díaz-Muñoz, Verónica M Rodríguez
ABSTRACT
Glyphosate (Glyph) is the active ingredient of several herbicide formulations. Reports of Glyph exposure in humans and animal models suggest that it may be neurotoxic. To evaluate the effects of Glyph on the nervous system, male Sprague-Dawley rats were given six intraperitoneal injections of 50, 100, or 150 mg Glyph/kg BW over 2 weeks (three injections/week). We assessed dopaminergic markers and their association with locomotor activity. Repeated exposure to Glyph caused hypoactivity immediately after each injection, and it was also apparent 2 days after the last injection in rats exposed to the highest dose. Glyph did not decrease monoamines, tyrosine hydroxylase (TH), or mesencephalic TH+ cells when measured 2 or 16 days after the last Glyph injection. In contrast, Glyph decreased specific binding to D1 dopamine (DA) receptors in the nucleus accumbens (NAcc) when measured 2 days after the last Glyph injection. Microdialysis experiments showed that a systemic injection of 150 mg Glyph/kg BW decreased basal extracellular DA levels and high-potassium-induced DA release in striatum. Glyph did not affect the extracellular concentrations of 3,4-dihydroxyphenylacetic acid or homovanillic acid. These results indicate that repeated Glyph exposure results in hypoactivity accompanied by decreases in specific binding to D1-DA receptors in the NAcc, and that acute exposure to Glyph has evident effects on striatal DA levels. Additional experiments are necessary in order to unveil the specific targets of Glyph on dopaminergic system, and whether Glyph could be affecting other neurotransmitter systems involved in motor control.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioUltra, for molecular biology, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
N-(Phosphonomethyl)glycine, 96%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Spiperone, solid
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
N-(Phosphonomethyl)glycine, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)