MilliporeSigma
  • TALEN-mediated generation and genetic correction of disease-specific human induced pluripotent stem cells.
TALEN-mediated generation and genetic correction of disease-specific human induced pluripotent stem cells.
Current gene therapy (2014-09-24)
Sivaprakash Ramalingam, Narayana Annaluru, Karthikeyan Kandavelou, Srinivasan Chandrasegaran
ABSTRACT
Generation and precise genetic correction of patient-derived hiPSCs have great potential in regenerative medicine. Such targeted genetic manipulations can now be achieved using gene-editing nucleases. Here, we report generation of cystic fibrosis (CF) and Gaucher's disease (GD) hiPSCs respectively from CF (homozygous for CFTRΔF508 mutation) and Type II GD [homozygous for β-glucocerebrosidase (GBA) 1448T>C mutation] patient fibroblasts, using CCR5- specific TALENs. Site-specific addition of loxP-flanked Oct4/Sox2/Klf4/Lin28/Nanog/eGFP gene cassette at the endogenous CCR5 site of patient-derived disease-specific primary fibroblasts induced reprogramming, giving rise to both monoallele (heterozygous) and biallele CCR5-modified hiPSCs. Subsequent excision of the donor cassette was done by treating CCR5-modified CF and GD hiPSCs with Cre. We also demonstrate site-specific correction of sickle cell disease (SCD) mutations at the endogenous HBB locus of patient-specific hiPSCs [TNC1 line that is homozygous for mutated β- globin alleles (βS/βS)], using HBB-specific TALENs. SCD-corrected hiPSC lines showed gene conversion of the mutated βS to the wild-type βA in one of the HBB alleles, while the other allele remained a mutant phenotype. After excision of the loxP-flanked DNA cassette from the SCD-corrected hiPSC lines using Cre, we obtained secondary heterozygous βS/βA hiPSCs, which express the wild-type (βA) transcript to 30-40% level as compared to uncorrected (βS/βS) SCD hiPSCs when differentiated into erythroid cells. Furthermore, we also show that TALEN-mediated generation and genetic correction of disease-specific hiPSCs did not induce any off-target mutations at closely related sites.
Product Description
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol solution, (Methanol:Dimethyl sulfoxide 1:1 (v/v))
Supelco
Methanol solution, contains 0.10 % (v/v) formic acid, UHPLC, suitable for mass spectrometry (MS), ≥99.5%
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ascorbic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Supelco
Linolenic acid, analytical standard
Sigma-Aldrich
Linolenic acid, ~70% (GC)
Supelco
Linoleic acid, analytical standard
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Linolenic acid, ≥99%
Sigma-Aldrich
Linoleic acid, ≥98%
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%