MilliporeSigma
  • The Arf GTPase-activating protein SMAP1 promotes transferrin receptor endocytosis and interacts with SMAP2.
The Arf GTPase-activating protein SMAP1 promotes transferrin receptor endocytosis and interacts with SMAP2.
Biochemical and biophysical research communications (2014-10-05)
Nobuhide Kobayashi, Shunsuke Kon, Yuji Henmi, Tomo Funaki, Masanobu Satake, Kenji Tanabe
ABSTRACT
Arf GTPase-activating proteins (Arf GAP) play important roles in the formation of the membrane vesicles that traffic between subcellular membranous organelles. The small Arf GTPase-activating protein (SMAP) subfamily of Arf GAPs has two members, SMAP1 and SMAP2, in mammals. The present study investigated whether these two proteins may have an overlapping function in addition to their previously reported distinct functions. Results showed that the presence of either SMAP1 or SMAP2 was sufficient for endocytosis of the transferrin receptor, and that transferrin incorporation was impaired only by the absence of both SMAP1 and SMAP2. This suggests the involvement of both SMAP1 and SMAP2 in transferrin endocytosis. Results also demonstrated a physical association between SMAP1 and SMAP2, which might serve as a basis for a functional interaction, and identified the intramolecular domains responsible for this association.
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O