MilliporeSigma
  • Laurate permeabilizes the paracellular pathway for small molecules in the intestinal epithelial cell model HT-29/B6 via opening the tight junctions by reversible relocation of claudin-5. [Corrected].
Laurate permeabilizes the paracellular pathway for small molecules in the intestinal epithelial cell model HT-29/B6 via opening the tight junctions by reversible relocation of claudin-5. [Corrected].
Pharmaceutical research (2014-03-19)
Isabel Dittmann, Maren Amasheh, Susanne M Krug, Alexander G Markov, Michael Fromm, Salah Amasheh
ABSTRACT
To mechanistically analyze effects of the medium-chain fatty acid laurate on transepithelial permeability in confluent monolayers of the intestinal epithelial cell line HT-29/B6, in context with an application as an absorption enhancer improving transepithelial drug permeation. Transepithelial resistance and apparent permeability for paracellular flux markers was measured using Ussing-type chambers. Two-path impedance spectroscopy was employed to differentiate between transcellular and paracellular resistance, and confocal imaging and Western blotting was performed. Laurate resulted in a substantial and reversible decrease in transepithelial resistance by 50% which was attributed to a decrease in paracellular resistance. Simultaneously, an increase in permeability for fluorescein (330 Da) was detected, while permeabilities for 4 kDa FITC-dextran and sulpho-NHS-SS-biotin (607 Da) remained unaltered. Confocal laser-scanning microscopy revealed a marked reduction of claudin-5, while other tight junction proteins including tricellulin, a protein preventing the paracellular passage of macromolecules, were not affected. Laurate induces an increase in paracellular permeability for molecules up to a molecular mass of 330 Da by retrieval of claudin-5 from tight junctions without affecting tricellular contacts and the paracellular passage of macromolecules. We hereby provide, for the first time, a mechanistical explanation of laurate-induced permeability enhancement on molecular level.
Sigma-Aldrich
Sodium dodecanoate, 99-100%