MilliporeSigma
Mesenchymal stem cells secrete immunologically active exosomes.
Stem cells and development (2013-12-26)
Bin Zhang, Yijun Yin, Ruenn Chai Lai, Soon Sim Tan, Andre Boon Hwa Choo, Sai Kiang Lim
ABSTRACT
Mesenchymal stem cells (MSCs) have been shown to secrete exosomes that are cardioprotective. Here, we demonstrated that MSC exosome, a secreted membrane vesicle, is immunologically active. MSC exosomes induced polymyxin-resistant, MYD88-dependent secreted embryonic alkaline phosphatase (SEAP) expression in a THP1-Xblue, a THP-1 reporter cell line with an NFκB-SEAP reporter gene. In contrast to lipopolysaccharide, they induced high levels of anti-inflammatory IL10 and TGFβ1 transcript at 3 and 72 h, and much attenuated levels of pro-inflammatory IL1B, IL6, TNFA and IL12P40 transcript at 3-h. The 3-h but not 72-h induction of cytokine transcript was abrogated by MyD88 deficiency. Primary human and mouse monocytes exhibited a similar exosome-induced cytokine transcript profile. Exosome-treated THP-1 but not MyD88-deficient THP-1 cells polarized activated CD4(+) T cells to CD4(+)CD25(+)FoxP3(+) regulatory T cells (Tregs) at a ratio of one exosome-treated THP-1 cell to 1,000 CD4(+) T cells. Infusion of MSC exosomes enhanced the survival of allogenic skin graft in mice and increased Tregs.
Sigma-Aldrich
Interleukin-6 human, Animal-component free, recombinant, expressed in E. coli, suitable for cell culture
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in HEK 293 cells, suitable for cell culture
Sigma-Aldrich
Interleukin-6 human, IL-6, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture