MilliporeSigma
DRR regulates AKT activation to drive brain cancer invasion.
Oncogene (2013-10-22)
A Dudley, M Sater, P U Le, G Trinh, M S Sadr, J Bergeron, G F Deleavey, B Bedell, M J Damha, K Petrecca
ABSTRACT
Glioblastoma (GBM) is the most common and invasive adult brain cancer. The rapid invasion of cancer cells into the normal brain is a major cause of treatment failure, yet the mechanisms that regulate this process are poorly understood. We have identified a novel mechanism of brain cancer invasion. We show that downregulated in renal cell carcinoma (DRR), which is newly expressed in invasive gliomas, recruits AKT to focal adhesions. This DRR- induced pathological relocalization of AKT bypasses commonly altered upstream signaling events and leads to AKT activation and invasion. We also developed an oligonucleotide therapeutic that reduces DRR expression and prevents glioma invasion in an in vivo preclinical model of the disease. Our findings identify DRR as a novel GBM target and show that oligonucleotides targeting DRR is a novel therapeutic approach for the treatment of DRR-positive GBMs.
Sigma-Aldrich
Epidermal Growth Factor, human, animal component free, EGF, recombinant, expressed in Escherichia coli, >97% (SDS-PAGE)
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Fluorouracil, European Pharmacopoeia (EP) Reference Standard
Supelco
Fluorouracil, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Fluorouracil, United States Pharmacopeia (USP) Reference Standard
Supelco
5-Fluorouracil, analytical standard