MilliporeSigma
  • Efficacy of raloxifene hydrochloride for the prevention of health care problems in patients who undergo surgery for endometrial cancer: a multicenter randomized clinical trial.
Efficacy of raloxifene hydrochloride for the prevention of health care problems in patients who undergo surgery for endometrial cancer: a multicenter randomized clinical trial.
International journal of gynecological cancer : official journal of the International Gynecological Cancer Society (2015-01-09)
Koji Nakamura, Kenjiro Sawada, Michiyo Sugiyama, Seiji Mabuchi, Takeshi Hisamatsu, Yukihiro Nishio, Kimihiko Ito, Tadashi Kimura, Shoji Kamiura, Ken-ichirou Morishige
ABSTRACT
Removal of the ovaries is common during surgery for endometrial cancer. However, because loss of the ovaries can cause several health problems in patients, strategies for the prevention of such problems need to be established. Hence, we decided to conduct a multicenter randomized clinical trial to assess the effect of raloxifene on bone mineral density (BMD), bone metabolism, and the lipid profile of patients who had undergone surgery for patients with endometrial cancer. Patients with endometrial cancer were enrolled after treatment. The participants were randomized into 2 groups: group 1 included 39 women who received alfacalcidol (1 μg/d) alone and group 2 included 37 women who received alfacalcidol and the test drug, raloxifene hydrochloride, at a dose of 60 mg/d. The BMD of lumbar spine and femoral neck, serum bone markers, as well as lipid profile parameters were evaluated at enrollment as well as 6, 12, and 24 months after the enrollment. The primary efficacy end point was the percentage change from baseline to 24 months in lumbar spine (L2-L4) and femoral neck BMD. Sixty-four women completed the 24-month study. At 24 months, the lumbar and femoral neck BMDs were significantly increased in group 2 compared with group 1 (3.5% vs -0.8% and 2.3% vs -2.8%, respectively). In group 2, low-density lipoprotein-cholesterol levels were significantly reduced by 13.6% and serum N-terminal telopeptide of type I collagen as well as bone-specific alkaline phosphatase values were significantly reduced by 16.7% and 25.7%, respectively. The patients who received adjuvant therapy for endometrial cancer showed a significantly higher response to raloxifene (5.8% vs 1.9%). Recurrence was detected in 2 (2.6%) patients in group 1. No severe adverse events were noted in any patient during the study period. Raloxifene exerts positive effects on BMD, bone metabolism, and lipid profile parameters and could provide an improved therapeutic option for patients with endometrial cancer.
Sigma-Aldrich
1α-Hydroxyvitamin D3, ≥97.0% (HPLC)
Alfacalcidol, European Pharmacopoeia (EP) Reference Standard