MilliporeSigma
  • Integrated metabolic flux and omics analysis of Synechocystis sp. PCC 6803 under mixotrophic and photoheterotrophic conditions.
Integrated metabolic flux and omics analysis of Synechocystis sp. PCC 6803 under mixotrophic and photoheterotrophic conditions.
Plant & cell physiology (2014-06-28)
Tsubasa Nakajima, Shuichi Kajihata, Katsunori Yoshikawa, Fumio Matsuda, Chikara Furusawa, Takashi Hirasawa, Hiroshi Shimizu
ABSTRACT
Cyanobacteria have flexible metabolic capability that enables them to adapt to various environments. To investigate their underlying metabolic regulation mechanisms, we performed an integrated analysis of metabolic flux using transcriptomic and metabolomic data of a cyanobacterium Synechocystis sp. PCC 6803, under mixotrophic and photoheterotrophic conditions. The integrated analysis indicated drastic metabolic flux changes, with much smaller changes in gene expression levels and metabolite concentrations between the conditions, suggesting that the flux change was not caused mainly by the expression levels of the corresponding genes. Under photoheterotrophic conditions, created by the addition of the photosynthesis inhibitor atrazine in mixotrophic conditions, the result of metabolic flux analysis indicated the significant repression of carbon fixation and the activation of the oxidative pentose phosphate pathway (PPP). Moreover, we observed gluconeogenic activity of upstream of glycolysis, which enhanced the flux of the oxidative PPP to compensate for NADPH depletion due to the inhibition of the light reaction of photosynthesis. 'Omics' data suggested that these changes were probably caused by the repression of the gap1 gene, which functions as a control valve in the metabolic network. Since metabolic flux is the outcome of a complicated interplay of cellular components, integrating metabolic flux with other 'omics' layers can identify metabolic changes and narrow down these regulatory mechanisms more effectively.
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, ≥95%
Supelco
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, LiChropur, ≥95.0% (GC)